CH6447916732 Chem Impex International, Inc.

O-Benzyl-L-threonine benzyl ester hydrochloride

  • MDL number MFCD11113160
  • Cas Number : 67580-86-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 107-112
Synonym L-Thr(Bzl)-OBzl HCl
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-o-i-benzyl-l-threonine-benzyl-ester-hydrochloride/6219?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-o-i-benzyl-l-threonine-benzyl-ester-hydrochloride/6219?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart