CH6447916738 Chem Impex International, Inc.

Fmoc-trans-4-aminocyclohexane acetic acid

  • MDL number MFCD04973171
  • Cas Number : 1217650-00-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 190-198
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-trans-4-aminocyclohexane-acetic-acid/6276?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-trans-4-aminocyclohexane-acetic-acid/6276?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart