CH6447916745 Chem Impex International, Inc.

Fmoc-N-(2-tert-butoxyethyl)glycine dicyclohexylammonium salt

  • MDL number MFCD08236869

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 113-118
Synonym Fmoc-Nhser(tBu)-OH DCHA
Purity 99
Storage temp. Store at 0-8
Physical Form White crystalline powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-i-n-i-2-i-tert-i-butoxyethyl-glycine-dicyclohexylammonium-salt/6477?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-i-n-i-2-i-tert-i-butoxyethyl-glycine-dicyclohexylammonium-salt/6477?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart