CH6447916751 Chem Impex International, Inc.

Boc-2-chloro-L-b-homophenylalanine

  • MDL number MFCD01076275
  • Cas Number : 218608-95-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 104-110
Synonym Boc-L-beta-HomoPhe(2-Cl)-OH
Purity 0.99
Storage temp. Store at 0-8
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-2-chloro-l-beta-homophenylalanine/6617?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-2-chloro-l-beta-homophenylalanine/6617?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart