CH6447916761 Chem Impex International, Inc.

Boc-3-methyl-D-beta-homophenylalanine

  • MDL number MFCD01860925
  • Cas Number : 269398-83-6
  • Molecular Weight : 293.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 96 - 102 °C
Purity 99.5
Storage temp. Store at 0 - 8 °C
Physical Form White to off-white powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-3-methyl-d-ß-homophenylalanine/6670?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-3-methyl-d-ß-homophenylalanine/6670?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart