CH6447916768 Chem Impex International, Inc.

Fmoc-(3-thienyl)-L-beta-homoalanine

  • MDL number MFCD01861085
  • Cas Number : 270263-01-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 156-162 °C
Purity 98
Storage temp. Store at 0-8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-3-thienyl-l-ß-homoalanine/6774?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-3-thienyl-l-ß-homoalanine/6774?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart