CH6447916774 Chem Impex International, Inc.

(S)-3-Amino-3-(4-hydroxyphenyl)propionic acid

  • MDL number MFCD04113697
  • Cas Number : 54732-46-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0-8 °C
Physical Form Off-white powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-s-i-3-amino-3-4-hydroxyphenyl-propionic-acid/6830?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-s-i-3-amino-3-4-hydroxyphenyl-propionic-acid/6830?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart