CH6447916787 Chem Impex International, Inc.

Boc-(R)-3-amino-3-(4-nitrophenyl)propionic acid

  • MDL number MFCD03427932
  • Cas Number : 500770-85-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 128-134 °C
Purity 99
Storage temp. Store at 0-8 °C
Physical Form White solid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-i-r-i-3-amino-3-4-nitrophenyl-propionic-acid/6983?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-i-r-i-3-amino-3-4-nitrophenyl-propionic-acid/6983?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart