CH6447916790 Chem Impex International, Inc.

Fmoc-(S)-3-amino-3-(3-methoxyphenyl)propionic acid

  • MDL number MFCD03427975
  • Cas Number : 501015-29-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Fmoc-L-beta-Phe(3-OMe)-OH
Purity 95
Storage temp. Store at 0-8
Physical Form White solid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-i-s-i-3-amino-3-3-methoxyphenyl-propionic-acid/7012?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-i-s-i-3-amino-3-3-methoxyphenyl-propionic-acid/7012?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart