CH6447916791 Chem Impex International, Inc.

Boc-(S)-3-amino-3-(2-hydroxyphenyl)propionic acid

  • MDL number MFCD03427901
  • Cas Number : 499995-78-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 152-158
Synonym Boc-L-beta-Phe(2-OH)-OH
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-i-s-i-3-amino-3-2-hydroxyphenyl-propionic-acid/7020?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-i-s-i-3-amino-3-2-hydroxyphenyl-propionic-acid/7020?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart