CH6447916796 Chem Impex International, Inc.

erythro-N-Boc-O-benzyl-L-tyrosine epoxide

  • MDL number MFCD04035566
  • Cas Number : 162536-84-7
  • Molecular Weight : 369.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-erythro-i-i-n-i-boc-i-o-i-benzyl-l-tyrosine-epoxide/7095?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-erythro-i-i-n-i-boc-i-o-i-benzyl-l-tyrosine-epoxide/7095?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart