CH6447916799 Chem Impex International, Inc.

Boc-(R)-alpha-phenethylproline

  • MDL number MFCD06659173
  • Cas Number : 1217805-48-5
  • Molecular Weight : 319.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 162-168 °C
Purity 99
Storage temp. Store at 0-8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-i-r-i-a-phenethylproline/7132?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-i-r-i-a-phenethylproline/7132?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart