CH6447916807 Chem Impex International, Inc.

(S)-a-(4-Biphenylmethyl)proline�

  • MDL number MFCD06659218
  • Cas Number : 1217722-65-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-s-i-a-4-biphenylmethyl-proline·hcl/7209?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-s-i-a-4-biphenylmethyl-proline·hcl/7209?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart