CH6447916816 Chem Impex International, Inc.

(S)-a-Propynylproline�

  • MDL number MFCD06659383
  • Cas Number : 1217828-88-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-s-i-a-propynylproline·hcl/7252?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-s-i-a-propynylproline·hcl/7252?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart