CH6447916825 Chem Impex International, Inc.

(R)-gamma-(1-Naphthalenylmethyl)-L-proline HCl

  • MDL number MFCD06659416
  • Cas Number : 1049740-26-2
  • Molecular Weight : 291.78

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 130-136 °C
Purity 98
Storage temp. Store at 0-8 °C
Physical Form Light brown powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-r-i-1-naphthalenylmethyl-l-proline·hcl/7286?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-r-i-1-naphthalenylmethyl-l-proline·hcl/7286?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart