CH6447916854 Chem Impex International, Inc.

(�rans-4-(3-Hydroxyphenyl)pyrrolidine-3-carboxylic acid hydrochloride

  • MDL number MFCD06659306
  • Cas Number : 1392213-98-7
  • Molecular Weight : 243.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0-8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/plusmn-i-trans-i-4-3-hydroxyphenyl-pyrrolidine-3-carboxylic-acid-hydrochloride/7401?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/plusmn-i-trans-i-4-3-hydroxyphenyl-pyrrolidine-3-carboxylic-acid-hydrochloride/7401?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart