CH6447916861 Chem Impex International, Inc.

2-(4-Boc-piperazinyl)-2-(3-bromophenyl)acetic acid

  • MDL number MFCD06659339
  • Cas Number : 885273-07-4
  • Molecular Weight : 399.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 171 - 172 °C
Purity 99
Storage temp. Store at 0 - 8 °C
Physical Form White solid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/2-4-boc-piperazinyl-2-3-bromophenyl-acetic-acid/7432?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/2-4-boc-piperazinyl-2-3-bromophenyl-acetic-acid/7432?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart