CH6447916864 Chem Impex International, Inc.

Nalpha-(4,4-Dimethyl-2,6-dioxocyclohex-1-ylidene)ethyl-Nbeta-allyloxycarbonyl-D-2,3-diamino-propionic acid dicyclohexylammonium salt

  • MDL number MFCD22493324
  • Molecular Weight : 533.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 126-132 °C
Purity 98
Storage temp. Store at 0-8 °C
Physical Form White crystalline powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-4-4-dimethyl-2-6-dioxocyclohex-1-ylidene-ethyl-i-n-i-sup-beta-sup-allyloxycarbonyl-d-2-3-diamino-propionic-acid-dicyclohexylammonium-salt/7515?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-4-4-dimethyl-2-6-dioxocyclohex-1-ylidene-ethyl-i-n-i-sup-beta-sup-allyloxycarbonyl-d-2-3-diamino-propionic-acid-dicyclohexylammonium-salt/7515?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart