CH6447916866 Chem Impex International, Inc.

Nalpha-1-(4,4-dimethyl-2,6-dioxocyclohex-1-ylidene)ethyl-Nepsilon-allyloxycarbonyl-L-lysine dicyclohexylammonium salt

  • MDL number MFCD08457691
  • Cas Number : 264230-73-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 0-8 °C
Physical Form White crystalline powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-1-4-4-dimethyl-2-6-dioxocyclohex-1-ylidene-ethyl-i-n-i-sup-epsilon-sup-allyloxycarbonyl-l-lysine-dicyclohexylammonium-salt/7521?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-1-4-4-dimethyl-2-6-dioxocyclohex-1-ylidene-ethyl-i-n-i-sup-epsilon-sup-allyloxycarbonyl-l-lysine-dicyclohexylammonium-salt/7521?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart