CH6447916870 Chem Impex International, Inc.

Nalpha-Z-Ngamma-allyloxycarbonyl-L-2,4-diaminobutyric acid dicyclohexylammonium salt

  • MDL number MFCD07366929
  • Molecular Weight : 517.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 120-126 °C
Purity 99
Storage temp. Store at 0-8 °C
Physical Form White crystalline powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-z-i-n-i-sup-gamma-sup-allyloxycarbonyl-l-2-4-diaminobutyric-acid-dicyclohexylammonium-salt/7592?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-z-i-n-i-sup-gamma-sup-allyloxycarbonyl-l-2-4-diaminobutyric-acid-dicyclohexylammonium-salt/7592?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart