CH6447916873 Chem Impex International, Inc.

Nalpha-Z-Nbeta-allyloxycarbonyl-D-2,3-diaminopropionic acid dicyclohexylammonium salt

  • MDL number MFCD08457692
  • Cas Number : 1423018-03-4
  • Molecular Weight : 503.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 169-175 °C
Purity 99
Storage temp. Store at 0-8 °C
Physical Form White crystalline powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-z-i-n-i-sup-beta-sup-allyloxycarbonyl-d-2-3-diaminopropionic-acid-dicyclohexylammonium-salt/7602?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-z-i-n-i-sup-beta-sup-allyloxycarbonyl-d-2-3-diaminopropionic-acid-dicyclohexylammonium-salt/7602?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart