CH6447916875 Chem Impex International, Inc.

L-b-Oleoyl-g-palmitoyl-a-lecithin

  • MDL number MFCD00077387
  • Cas Number : 26853-31-6
  • Molecular Weight : 760.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 0-8°C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/l-b-oleoyl-g-palmitoyl-a-lecithin/7620?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/l-b-oleoyl-g-palmitoyl-a-lecithin/7620?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart