CH6447916917 Chem Impex International, Inc.

2-Amino-3-pyridinecarboxaldehyde

  • MDL number MFCD03453200
  • Cas Number : 7521-41-7
  • Molecular Weight : 158.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 98 - 104 °C
Purity 99
Storage temp. Store at 0 - 8 °C
Physical Form Yellow crystalline powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/2-amino-3-pyridinecarboxaldehyde·hcl/9046?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/2-amino-3-pyridinecarboxaldehyde·hcl/9046?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart