CH6447916924 Chem Impex International, Inc.

5-Methoxy-2,3-dihydro-1H-indole hydrochloride

  • MDL number MFCD07783777

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 5-Methoxyindoline hydrochloride
Purity 96
Storage temp. Store at 0-8
Physical Form Light brown to brown powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/5-methoxy-2-3-dihydro-1h-indole-hydrochloride/9191?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/5-methoxy-2-3-dihydro-1h-indole-hydrochloride/9191?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart