CH6447916928 Chem Impex International, Inc.

8-Boc-3,8-diaza-bicyclo[3.2.1]octane

  • MDL number MFCD17016708
  • Cas Number : 149771-44-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 97-103 �C
Purity 95
Storage temp. Store at 0-8
Physical Form White crystalline powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/8-boc-3-8-diaza-bicyclo-3-2-1-octane/9237?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/8-boc-3-8-diaza-bicyclo-3-2-1-octane/9237?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart