CH6447916988 Chem Impex International, Inc.

1-[3-(2-Methoxy-ethoxy)phenyl]piperazine dihydrochloride

  • MDL number MFCD09701325

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Store at 0-8°C
Physical Form Tan solid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/1-3-2-methoxy-ethoxy-phenyl-piperazine-dihydrochloride/10814?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/1-3-2-methoxy-ethoxy-phenyl-piperazine-dihydrochloride/10814?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart