CH6447917005 Chem Impex International, Inc.

5-Methoxy-2,3-dihydro-1H-isoindole hydrochloride

  • MDL number MFCD09997806
  • Cas Number : 1159822-61-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 5-Methoxyisoindoline hydrochloride
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/5-methoxy-2-3-dihydro-1-i-h-i-isoindole-hydrochloride/11477?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/5-methoxy-2-3-dihydro-1-i-h-i-isoindole-hydrochloride/11477?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart