CH6447917015 Chem Impex International, Inc.

6-Chloro-D-tryptophan

  • MDL number MFCD06202361
  • Cas Number : 56632-86-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8 °C
Physical Form Pale yellow powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/6-chloro-d-tryptophan/12100?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/6-chloro-d-tryptophan/12100?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart