CH6447917110 Chem Impex International, Inc.

(+)-Biotin-PFP-ester

  • MDL number MFCD09952636
  • Cas Number : 120550-35-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 195
Purity 98
Storage temp. Store at 0-8
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/biotin-pfp-ester/13625?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/biotin-pfp-ester/13625?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart