CH6447917227 Chem Impex International, Inc.

(S)-1,2,3,4-Tetrahydronaphthoic acid

  • MDL number MFCD07368365
  • Cas Number : 85977-52-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 53-59
Purity 98
Storage temp. Store at 0-8
Physical Form White solid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/s-1-2-3-4-tetrahydronaphthoic-acid/13751?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/s-1-2-3-4-tetrahydronaphthoic-acid/13751?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart