CH6447917258 Chem Impex International, Inc.

S-(4-Chlorobenzyl)Isothiouronium chloride

  • MDL number MFCD00012584
  • Cas Number : 544-47-8
  • Molecular Weight : 237.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 202-209
Purity 99
Storage temp. Store at 0-8
Physical Form White solid
Shipping Temperature RT
Country of Origin US
Product Link http://www.chemimpex.com/product/productinfo/i-s-i-4-chlorobenzyl-isothiouronium-chloride/13785?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-s-i-4-chlorobenzyl-isothiouronium-chloride/13785?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart