CH6447917427 Chem Impex International, Inc.

2-(4-Fluorophenyl)-2-phenylacetic acid

  • MDL number MFCD01631958
  • Cas Number : 723-69-3
  • Molecular Weight : 230.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Store at 0-8
Physical Form Light yellow powder
Shipping Temperature RT
Country of Origin US
Product Link http://www.chemimpex.com/product/productinfo/2-4-fluorophenyl-2-phenylacetic-acid/13955?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/2-4-fluorophenyl-2-phenylacetic-acid/13955?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart