CH6447917494 Chem Impex International, Inc.

[5-(4-Chlorophenyl)-2-thienyl]methylamine hydrochloride

  • MDL number MFCD02089432
  • Cas Number : 1166853-02-6
  • Molecular Weight : 260.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 93
Storage temp. Store at 0-8
Physical Form Off-white solid
Shipping Temperature RT
Country of Origin US
Product Link http://www.chemimpex.com/product/productinfo/5-4-chlorophenyl-2-thienyl-methylamine-hydrochloride/14022?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/5-4-chlorophenyl-2-thienyl-methylamine-hydrochloride/14022?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart