CH6447917721 Chem Impex International, Inc.

Methyl 3-oxo-3-(thiophen-2-yl)propanoate

  • MDL number MFCD03424761
  • Cas Number : 134568-16-4
  • Molecular Weight : 184.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0-8
Physical Form Yellow color liquid
Shipping Temperature RT
Country of Origin US
Boiling point 125-128 �mm Hg
Product Link http://www.chemimpex.com/product/productinfo/methyl-3-oxo-3-thiophen-2-yl-propanoate/18847?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/methyl-3-oxo-3-thiophen-2-yl-propanoate/18847?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart