CH6447917736 Chem Impex International, Inc.

4-Nitro-3-picoline

  • MDL number MFCD03095084
  • Cas Number : 1678-53-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 26-32
Synonym 4-Nitro-3-methylpyridine
Purity 96
Storage temp. Store at 0-8
Physical Form Pale yellow crystalline powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/4-nitro-3-picoline/18936?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/4-nitro-3-picoline/18936?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart