CH6447917777 Chem Impex International, Inc.

2-Deoxy-L-ribose

  • MDL number MFCD00132941
  • Cas Number : 18546-37-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0-8
Physical Form White to off-white crystalline powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/2-deoxy-l-ribose/19269?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/2-deoxy-l-ribose/19269?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart