CH6447917795 Chem Impex International, Inc.

1,7-Bis-Boc-1,4,7-triazaheptane

  • MDL number MFCD11226825
  • Cas Number : 117499-16-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8
Physical Form Syrupy liquid to white solid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/1-7-bis-boc-1-4-7-triazaheptane/19327?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/1-7-bis-boc-1-4-7-triazaheptane/19327?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart