CH6447917871 Chem Impex International, Inc.

(4S)-3-[5-(4-Fluorophenyl)-1,5-dioxopenyl]-4-phenyl-2-oxazolidinone

  • MDL number MFCD08061377
  • Cas Number : 189028-93-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0-8
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/4-i-s-i-3-5-4-fluorophenyl-1-5-dioxopenyl-4-phenyl-2-oxazolidinone/19414?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/4-i-s-i-3-5-4-fluorophenyl-1-5-dioxopenyl-4-phenyl-2-oxazolidinone/19414?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart