CH6447918013 Chem Impex International, Inc.

Fmoc-L-Lys(Boc)(2,3:4,5-di-O-isopropylidene-1-deoxyfructopyranosyl)-OH

  • MDL number MFCD26793606
  • Molecular Weight : 710.82

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 79-83 �C
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature RT
Country of Origin US
Product Link http://www.chemimpex.com/product/productinfo/fmoc-l-lys-boc-2-3-4-5-di-o-isopropylidene-1-deoxyfructopyranosyl-oh/19613?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-l-lys-boc-2-3-4-5-di-o-isopropylidene-1-deoxyfructopyranosyl-oh/19613?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart