CH6447918174 Chem Impex International, Inc.

Adenine phosphate salt

  • MDL number MFCD00035170
  • Cas Number : 70700-30-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 6-Aminopurine phosphate salt
Purity 99
Storage temp. Store at 0-8 �C
Physical Form White or almost white crystalline powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/adenine-phosphate-salt/19757?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/adenine-phosphate-salt/19757?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart