CH6447918231 Chem Impex International, Inc.

Potassium bromate

  • MDL number MFCD00011359
  • Cas Number : 7758-01-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 434 �C (Approx.); begins to decompose at 370 �C.
Purity 99.8
Storage temp. Store at RT
Physical Form White crystalline powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/potassium-bromate/20011?cid=0

Safety Information

hcodes 5.1
MSDS http://www.chemimpex.com/product/productinfo/potassium-bromate/20011?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart