CH6447918285 Chem Impex International, Inc.

(-)-Di-p-anisoyl-L-tartaric Acid

  • MDL number MFCD02682986
  • Cas Number : 50583-51-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at RT
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/di-p-anisoyl-l-tartaric-acid/20161?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/di-p-anisoyl-l-tartaric-acid/20161?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart