CH6447918286 Chem Impex International, Inc.

(+)-Di-p-anisoyl-D-tartaric Acid

  • MDL number MFCD07368366
  • Cas Number : 191605-10-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at RT
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/di-p-anisoyl-d-tartaric-acid/20162?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/di-p-anisoyl-d-tartaric-acid/20162?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart