CH6458423817 Chem Impex International, Inc.

Acetyl-L-cystine bis-methyl ester

  • MDL number MFCD00083613
  • Cas Number : 32381-28-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (Ac-L-Cys-OMe)2
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/acetyl-l-cystine-bis-methyl-ester-br-disulfide-bond/2984?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/acetyl-l-cystine-bis-methyl-ester-br-disulfide-bond/2984?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart