CH6458423818 Chem Impex International, Inc.

4-Nitro-DL-phenylalanine hydrate

  • MDL number MFCD00149511

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym DL-Phe(4-NO2)-OH�
Purity 99
Storage temp. Store at 0-8
Physical Form Faintly yellow powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/4-nitro-dl-phenylalanine-hydrate/3146?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/4-nitro-dl-phenylalanine-hydrate/3146?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart