CH6458423823 Chem Impex International, Inc.

L-Aspartic acid alpha-4-nitroanilide hydrochloride

  • MDL number MFCD26793471
  • Cas Number : 154564-03-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 176-181 °C
Purity 99
Storage temp. Store at 0-8 °C
Physical Form Light yellow crystalline powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/l-aspartic-acid-a-4-nitroanilide/3864?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/l-aspartic-acid-a-4-nitroanilide/3864?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart