CH6458423824 Chem Impex International, Inc.

L-Aspartic acid b-naphthylamide

  • MDL number MFCD00037739
  • Cas Number : 635-91-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym L-Asp-betaNA
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/l-aspartic-acid-ß-naphthylamide/3865?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/l-aspartic-acid-ß-naphthylamide/3865?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart