CH6458423826 Chem Impex International, Inc.

N-Methyl-DL-valine

  • MDL number MFCD00069559
  • Cas Number : 2566-32-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 0-8°C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-methyl-dl-valine-hydrochloride/4175?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-methyl-dl-valine-hydrochloride/4175?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart