CH6458423831 Chem Impex International, Inc.

N-omega-Hydroxy-L-norarginine diacetate salt

  • MDL number MFCD03453555
  • Cas Number : 189302-40-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at = -4 °C
Physical Form Off-white powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/n-w-hydroxy-l-norarginine-diacetate-salt/4524?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/n-w-hydroxy-l-norarginine-diacetate-salt/4524?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart