CH6458423837 Chem Impex International, Inc.

b-Cyclohexyl-L-alanine amide

  • MDL number MFCD21606314
  • Cas Number : 145232-34-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 252-257 �C
Synonym L-Cha-NH2 HCl
Purity 98
Storage temp. Store at 0-8
Physical Form White to off-white solid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/beta-cyclohexyl-l-alanine-amide-hydrochloride/6059?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/beta-cyclohexyl-l-alanine-amide-hydrochloride/6059?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart